Niobium(V) chloride ≥99.99% (Nb basis) (20-200ppm Ta), PURATREM
Supplier: STREM CHEMICALS INC MS
|
Danger
|
41-1250-25G
41-1250-5G
41-1250-100G
100205-114EA
157.11
USD
100205-114
100205-116
100200-680
Niobium(V) chloride ≥99.99% (Nb basis) (20-200ppm Ta), PURATREM
Niobium(V) chloride
Formula:
Cl₅Nb Boiling Pt: 254 °C Melting Pt: 204…206 °C Density: 2.75 g/cm³ (20 °C) |
MDL Number:
MFCD00011127 CAS Number: 10026-12-7 EINECS: 233-059-8 UN: 3260 ADR: 8,II Merck Index: 12,06649 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...