Zirconium tetrachloride ≥99.95% (Zr basis), sublimated

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Zirconium (IV) chloride

40-4046-50G 40-4046-250G 40-4046-1000G
100207-670EA 83.51 USD
100207-670 100207-668 100196-206
Zirconium tetrachloride ≥99.95% (Zr basis), sublimated
Zirconium tetrachloride
Formula: Cl₄Zr
MW: 233.03 g/mol
Boiling Pt: 3577 (subl.)
Melting Pt: 437
Density: 2.803
Storage Temperature: Ambient
MDL Number: MFCD00011306
CAS Number: 10026-11-6
EINECS: 233-058-2
UN: 2503
ADR: 8 III
Merck Index: 12,10307

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR