Zirconyl chloride hydrate ≥99.99% (Zr basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Zirconium(IV) dichloride oxide hydrate, Zirconium dichloride oxide hydrate

40-2500-100G 40-2500-25G
100199-874EA 598.71 USD
100199-874 100211-078
Zirconyl chloride hydrate ≥99.99% (Zr basis), PURATREM
Zirconyl chloride hydrate
Formula: Cl₂OZr·H₂O
MW: 196.14
MDL Number: MFCD00149897
CAS Number: 15461-27-5
EINECS: 231-717-9
UN: 1759
ADR: 8 III
Merck Index: 14,10184

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR