Zirconium(IV) tert-butanolate ≥99.99% (Zr basis), PURATREM
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Zirconium(IV) t-butoxide, Zirconium (IV) tert-butoxide, Zirconium (IV) tert-butylate
40-1750-2G
40-1750-10G
40-1750-50G
100206-600EA
93.42
USD
100206-600
100206-598
100200-270
Zirconium(IV) tert-butanolate ≥99.99% (Zr basis), PURATREM
Zirconium(IV) tert-butanolate
Alkoxide, Metal Alkoxides, Volatile Precursors for CVD
Formula:
C₁₆H₃₆O₄Zr MW: 383.68 g/mol Boiling Pt: 90 °C (5 mmHg) Density: 0.96 Flash Pt: 185 °F |
MDL Number:
MFCD00075085 CAS Number: 2081-12-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...