Zirconium(IV) tert-butanolate ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Zirconium(IV) t-butoxide, Zirconium (IV) tert-butoxide, Zirconium (IV) tert-butylate

40-1749-5G 40-1749-25G 40-1749-100G
100203-780EA 141.54 USD
100203-780 100207-398 100207-666
Zirconium(IV) tert-butanolate ≥99%
Zirconium(IV) tert-butanolate

Alkoxide, Metal Alkoxides, Volatile Precursors for CVD

Formula: C₁₆H₃₆O₄Zr
MW: 383.68 g/mol
Boiling Pt: 90 °C (5 mmHg)
Density: 0,96
Flash Pt: 185 °F
MDL Number: MFCD00075085
CAS Number: 2081-12-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR