Zirconium ≥99.5%, crystal bar turning

Supplier: STREM CHEMICALS INC MS

40-0055-10G 40-0055-50G
100199-388EA 164.18 USD
100199-388 100203-776
Zirconium ≥99.5%, crystal bar turning
Zirconium
Formula: Zr
MW: 91.22 g/mol
Boiling Pt: 3577 °C
Melting Pt: 1852 °C
Density: 6.506
MDL Number: MFCD00011303
CAS Number: 7440-67-7
EINECS: 231-176-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR