Strontium oxide ≥99.9%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
38-2250-10G
38-2250-50G
100203-106EA
151.45
USD
100203-106
100200-676
Strontium oxide ≥99.9%
Strontium oxide
Formula:
SrO Boiling Pt: 3000 °C (1013 hPa) Melting Pt: 2430 °C Density: 4.7 g/cm³ (20 °C) |
MDL Number:
MFCD00049558 CAS Number: 1314-11-0 EINECS: 215-219-9 UN: 3262 ADR: 8,II Merck Index: 12,09009 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...