Benzeneselenenyl chloride
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Phenylselenium chloride, Phenylselenenyl chloride, Chloroselenobenzene, Phenylselenyl chloride
34-1400-10G
34-1400-2G
100210-638EA
309.97
USD
100210-638
100201-598
Benzeneselenenyl chloride
Benzeneselenenyl chloride
Formula:
(C₆H₅)SeCl MW: 191.52 g/mol Boiling Pt: 237 Melting Pt: 64…65 °C |
MDL Number:
MFCD00000478 CAS Number: 5707-04-0 EINECS: 227-196-2 UN: 2923 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...