Diphenyl selenide
Supplier: STREM CHEMICALS INC MS
|
Danger
|
34-0700-10G
34-0700-2G
100197-112EA
627.02
USD
100197-112
100197-114
Diphenyl selenide
Diphenyl selenide
Formula:
C₁₂H₁₀Se Boiling Pt: 301 - 302 °C (1013 hPa) Melting Pt: 61 - 63 °C Density: 1.33 g/cm³ (15 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00014067 CAS Number: 1132-39-4 EINECS: 214-474-3 UN: 3440 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...