Dimethyl selenide ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

34-0550-10G 34-0550-50G
100199-856EA 1464.92 USD
100199-856 100199-858
Dimethyl selenide ≥99%
Dimethyl selenide

Metal Alkyls, Volatile Precursors for CVD

Formula: C₂H₆Se
MW: 109.03 g/mol
Boiling Pt: 57
Density: 1.4077
Storage Temperature: Refrigerator
MDL Number: MFCD00014848
CAS Number: 593-79-3
EINECS: 209-807-4
UN: 3440
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR