Selenious acid ≥99.999% (Se basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Danger

34-0100-100G 34-0100-25G
100203-100EA 976.62 USD
100203-100 100207-388
Selenious acid ≥99.999% (Se basis), PURATREM
Selenious acid
Formula: H₂O₃Se
Boiling Pt: 315 °C (1013 hPa)
Melting Pt: 70 °C (dec.)
Density: 1.998 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011331
CAS Number: 7783-00-8
EINECS: 231-974-7
UN: 3283
ADR: 6.1,II
Merck Index: 14,08430

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR