Tetraphenylarsonium chloride monohydrate ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Tetraphenylarsonium(V) chloride hydrate

33-3050-1G 33-3050-25G 33-3050-5G
100207-658EA 123.14 USD
100207-658 100200-926 100200-928
Tetraphenylarsonium chloride monohydrate ≥99%
Tetraphenylarsonium chloride monohydrate
Formula: C₂₄H₂₀AsCl·1H₂O
MW: 436.81 g/mol
Melting Pt: 260 °C
MDL Number: MFCD00011908
CAS Number: 104170-16-3
EINECS: 208-070-6
UN: 1557
ADR: 6.1,II
Merck Index: 12,09374

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR