Lithium chloride monohydrate ≥99.996% (Li basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Lithium chloride hydrate, Hydrochloric acid lithium salt monohydrate

03-0900-50G 03-0900-10G
100201-304EA 397.72 USD
100201-304 100203-886
Lithium chloride monohydrate ≥99.996% (Li basis), PURATREM
Lithium chloride monohydrate
Formula: ClLi·1H₂O
MW: 60.41 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00149764
CAS Number: 16712-20-2
EINECS: 231-212-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR