Zinc ≥99.999%, shot
Supplier: STREM CHEMICALS INC MS
|
Warning
|
30-0070-1000G
30-0070-50G
30-0070-250G
100204-030EA
1321.97
USD
100204-030
100198-172
100198-170
Zinc ≥99.999%, shot
Zinc
Formula:
Zn MW: 65.39 g/mol Boiling Pt: 908 Melting Pt: 420 Density: 7.14 Storage Temperature: Ambient |
MDL Number:
MFCD00011291 CAS Number: 7440-66-6 EINECS: 231-175-3 Merck Index: 12,10255 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...