Zinc ≥99.9999%, rods
Supplier: STREM CHEMICALS INC MS
30-0060-50MM
30-0060-250MM
100201-206EA
321.29
USD
100201-206
100198-740
Zinc ≥99.9999%, rods
Zinc
Formula:
Zn Boiling Pt: 907 °C Melting Pt: 420 °C Density: 7.14 Storage Temperature: Ambient |
MDL Number:
MFCD00011291 CAS Number: 7440-66-6 EINECS: 231-175-3 Merck Index: 12,10255 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...