Zinc ≥99.98%, foil
Supplier: STREM CHEMICALS INC MS
30-0047-250CM
30-0047-50CM
100209-896EA
358.09
USD
100209-896
100203-086
Zinc ≥99.98%, foil
Zinc
Formula:
Zn Boiling Pt: 907 °C Melting Pt: 420 °C Density: 7.14 Storage Temperature: Ambient |
MDL Number:
MFCD00011291 CAS Number: 7440-66-6 EINECS: 231-175-3 Merck Index: 12,10255 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...