Copper(I) iodide ≥99.999% (Cu basis), PURATREM
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Cuprous iodide
29-0485-50G
29-0485-10G
100211-060EA
311.38
USD
100211-060
100211-058
Copper(I) iodide ≥99.999% (Cu basis), PURATREM
Copper(I) iodide
Formula:
CuI MW: 190.45 g/mol Boiling Pt: 1290 °C (1013 hPa) Melting Pt: 602 °C Density: 5.62 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010978 CAS Number: 7681-65-4 EINECS: 231-674-6 UN: 3077 ADR: 9,III Merck Index: 13,02692 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...