Copper ≥99.9999%, shot
Supplier: STREM CHEMICALS INC MS
29-0085-10G
29-0085-50G
100201-576EA
89.17
USD
100201-576
100201-578
Copper ≥99.9999%, shot
Copper
Formula:
Cu Boiling Pt: 2595 °C (1013 hPa) Melting Pt: 1083 °C Density: 8.92 Storage Temperature: Ambient |
MDL Number:
MFCD00010965 CAS Number: 7440-50-8 EINECS: 231-159-6 Merck Index: 13,02545 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...