Copper ≥99.9%, spherical powder

Supplier: STREM CHEMICALS INC MS
Danger

29-0075-250G 29-0075-1000G
100200-452EA 90.58 USD
100200-452 100203-084
Copper ≥99.9%, spherical powder
Copper
Formula: Cu
Boiling Pt: 2567 °C (1013 hPa)
Melting Pt: 1083 °C
Density: 8,92
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
UN: 3089
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR