Nickel (II) oxide ≥99.998% (Ni basis), PURATREM
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Nickel monoxide, Nickel oxide, Bunsenite
28-1475-50G
28-1475-10G
100206-104EA
515.2
USD
100206-104
100204-608
Nickel (II) oxide ≥99.998% (Ni basis), PURATREM
Nickel (II) oxide
Formula:
NiO Melting Pt: 1990 °C Density: 4.6 to 6.67 g/cm³ (20 °C) |
MDL Number:
MFCD00011145 CAS Number: 1313-99-1 EINECS: 215-215-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...