Nickel(II) acetylacetonate, anhydrous ≥95%
Supplier: STREM CHEMICALS INC MS
Synonyms:
Bis(2,4-pentanedionato-O,O')nickel (II), Nickel (II) bis(2,4-pentanedionato-O,O'), Nickel acetylacetonate, Nickel (II) 2,4-pentanedionate
28-1130-100G
28-1130-25G
100198-732EA
363.75
USD
100198-732
100209-882
Nickel(II) acetylacetonate, anhydrous ≥95%
Nickel(II) acetylacetonate
Metal Beta-diketonates, Volatile Metal Carbonyls for CVD & ALD, Volatile Precursors for CVD
Formula:
C₁₀H₁₄NiO₄ MW: 256.91 g/mol Boiling Pt: 220 Melting Pt: 208 (dec.) Density: 1.455 g/cm³ (20 °C) |
MDL Number:
MFCD00000024 CAS Number: 3264-82-2 EINECS: 221-875-7 Merck Index: 12,06584 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...