Nickel ≥99.9%, pellets
Supplier: STREM CHEMICALS INC MS
|
Warning
|
28-0005-1000G
28-0005-250G
100210-636EA
343.94
USD
100210-636
100200-446
Nickel ≥99.9%, pellets
Nickel
Formula:
Ni Boiling Pt: 2732 °C Melting Pt: 1453 °C Density: 8.9 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011137 CAS Number: 7440-02-0 EINECS: 231-111-4 UN: 3089 ADR: 4.1,III Merck Index: 12,06582 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...