Cobalt ≥99.9%, pieces
Supplier: STREM CHEMICALS INC MS
|
Danger
|
27-0300-100G
27-0300-500G
100202-656EA
125.97
USD
100202-656
100209-478
Cobalt ≥99.9%, pieces
Cobalt
Formula:
Co MW: 58.93 g/mol Boiling Pt: 2870 Melting Pt: 1495 Density: 8.9 Storage Temperature: Ambient |
MDL Number:
MFCD00010935 CAS Number: 7440-48-4 EINECS: 231-158-0 Merck Index: 12,02488 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...