Cobalt ≥99.9%, foil
Supplier: STREM CHEMICALS INC MS
|
Danger
|
27-0200-1PC
27-0200-5PC
100200-916EA
186.83
USD
100200-916
100200-918
Cobalt ≥99.9%, foil
Cobalt
Formula:
Co Boiling Pt: 2870 °C Melting Pt: 1495 °C Density: 8.9 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010935 CAS Number: 7440-48-4 EINECS: 231-158-0 Merck Index: 12,02488 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...