Cobalt ≥99.99%, powder

Supplier: STREM CHEMICALS INC MS
Danger

27-0045-10G 27-0045-50G
100200-912EA 165.6 USD
100200-912 100200-914
Cobalt ≥99.99%, powder
Cobalt
Formula: Co
Boiling Pt: 2870 °C
Melting Pt: 1495 °C
Density: 8.9
MDL Number: MFCD00010935
CAS Number: 7440-48-4
EINECS: 231-158-0
UN: 3089
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR