Cobalt ≥99.8%, powder

Supplier: STREM CHEMICALS INC MS
Danger

27-0040-250G 27-0040-50G
100203-066EA 363.75 USD
100203-066 100205-058
Cobalt ≥99.8%, powder
Cobalt
Formula: Co
MW: 58.93 g/mol
Boiling Pt: 2870
Melting Pt: 1495
Density: 8.9
MDL Number: MFCD00010935
CAS Number: 7440-48-4
EINECS: 231-158-0
UN: 3089
ADR: 4.1 II
Merck Index: 12,02488

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR