Iron(III) oxide 99,995 Fe basis, PURATREM

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Ferric oxide, Ferric Oxide, (iron(III) oxide), Diiron trioxide

26-2750-10G 26-2750-50G
100196-182EA 93.42 USD
100196-182 100201-558
Iron(III) oxide 99,995 Fe basis, PURATREM
Iron(III) oxide
Formula: Fe₂O₃
MW: 159.69 g/mol
Melting Pt: 1538
Density: 5.24 g/cm³ (20 °C)
MDL Number: MFCD00011008
CAS Number: 1309-37-1
EINECS: 215-168-2
Merck Index: 13,04058

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR