Iron(III) acetylacetonate 99

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Tris(2,4-pentanedionato-O,O')iron (III), Ferric acetylacetonate, Iron (III) 2,4-pentanedionate, Iron (III) tris(2,4-pentanedionato-O,O')

26-2300-250G 26-2300-50G
100199-838EA 131.63 USD
100199-838 100201-556
Iron(III) acetylacetonate 99
Iron(III) acetylacetonate
Formula: C₁₅H₂₁FeO₆
MW: 353.18 g/mol
Boiling Pt: 100
Melting Pt: 184 °C
Density: 5.24
MDL Number: MFCD00000020
CAS Number: 14024-18-1
EINECS: 237-853-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR