Ferrocene ≥99%

Supplier: STREM CHEMICALS INC MS

26-1700-100G 26-1700-500G 26-1700-2000G
100209-858EA 62.28 USD
100209-858 100211-048 100203-758
Ferrocene ≥99%
Ferrocene

Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD

Formula: C₁₀H₁₀Fe
MW: 186.04 g/mol
Boiling Pt: 249
Melting Pt: 172…173 °C
Density: 1.49 g/cm³ (20 °C)
MDL Number: MFCD00001427
CAS Number: 102-54-5
EINECS: 203-039-3
UN: 1325
ADR: 4.1 II
Merck Index: 13,04070

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR