Acetylferrocene ≥99.5%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
26-0050-100G
26-0050-5G
26-0050-25G
100196-924EA
680.8
USD
100196-924
100204-014
100200-906
Acetylferrocene ≥99.5%
Acetylferrocene
Formula:
C₁₂H₁₂FeO Melting Pt: 83 °C Density: 1 g/cm³ (20 °C) |
MDL Number:
MFCD00001432 CAS Number: 1271-55-2 EINECS: 215-043-2 UN: 2811 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...