Manganese(IV) oxide ≥99.995% (Mn basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Manganese dioxide, Manganese(IV) oxide, activated, Manganese peroxide, Mangan(IV)-oxid

25-1380-10G 25-1380-50G
100196-168EA 108.98 USD
100196-168 100195-910
Manganese(IV) oxide ≥99.995% (Mn basis), PURATREM
Manganese(IV) oxide
Formula: MnO₂
MW: 86.94 g/mol
Melting Pt: 535 °C (dec.)
Density: 5.03 g/cm³ (20 °C)
MDL Number: MFCD00003463
CAS Number: 1313-13-9
EINECS: 215-202-6
UN: 1479
ADR: 5.1,II
Merck Index: 14,05730

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR