Manganese(IV) oxide

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Manganese dioxide, Manganese(IV) oxide, activated, Manganese peroxide, Mangan(IV)-oxid

25-1360-50G 25-1360-250G
100208-392EA 58.03 USD
100208-392 100205-028
Manganese(IV) oxide
Manganese(IV) oxide
Formula: MnO₂
MW: 86.94 g/mol
Melting Pt: 535 °C
Density: 5.03 g/cm³ (20 °C)
MDL Number: MFCD00003463
CAS Number: 1313-13-9
EINECS: 215-202-6
UN: 1479
ADR: 5.1,II
Merck Index: 14,05730

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR