Manganese(III) acetylacetonate ≥90%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Manganese (III) tris(2,4-pentanedionato-O,O'), Tris(2,4-pentanedionato-O,O')manganese (III), Manganese (III) 2,4-pentanedionate
25-1000-100G
25-1000-25G
100197-102EA
317.05
USD
100197-102
100206-496
Manganese(III) acetylacetonate ≥90%
Manganese(III) acetylacetonate
Formula:
C₁₅H₂₁MnO₆ MW: 352.27 g/mol Melting Pt: 160 |
MDL Number:
MFCD00000023 CAS Number: 14284-89-0 EINECS: 238-188-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...