Manganese(III) acetate dihydrate ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Acetic acid manganese (III) salt dihydrate, Manganese triacetate dihydrate
25-0750-25G
25-0750-5G
100209-794EA
232.12
USD
100209-794
100209-796
Manganese(III) acetate dihydrate ≥98%
Manganese(III) acetate dihydrate
Formula:
(H₃CCOO)₂Mn·2H₂O MW: 209.06 g/mol |
MDL Number:
MFCD00150022 CAS Number: 19513-05-4 EINECS: 213-602-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...