Chromium hexacarbonyl ≥99%, sublimated
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Chromium carbonyl, Hexacarbonylchromium, Hexacarbonylchromium(0), Chromium(0) hexacarbonyl
24-0180-5G
24-0180-500G
24-0180-100G
100196-154EA
89.17
USD
100196-154
100196-152
100207-376
100207-374
Chromium hexacarbonyl ≥99%, sublimated
Chromium hexacarbonyl
Metal Carbonyls, Volatile Metal Carbonyls for CVD & ALD, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD
Formula:
C₆CrO₆ MW: 220.06 g/mol Boiling Pt: 210 °C (1013 hPa) Melting Pt: 90 °C Density: 1.77 g/cm³ (20 °C) |
MDL Number:
MFCD00010945 CAS Number: 13007-92-6 EINECS: 235-852-4 UN: 3466 ADR: 6.1,II Merck Index: 13,02253 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...