Chromium ≥99.997%, pellets

Supplier: STREM CHEMICALS INC MS

24-0040-25G 24-0040-100G
100209-786EA 94.83 USD
100209-786 100198-708
Chromium ≥99.997%, pellets
Chromium
Formula: Cr
MW: 52
Boiling Pt: 2469
Melting Pt: 1840
Density: 7.20
Storage Temperature: Ambient
MDL Number: MFCD00010944
CAS Number: 7440-47-3
EINECS: 231-157-5
Merck Index: 12,02288

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR