Vanadium(III) acetylacetonate ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

23-2250-25G 23-2250-100G
100208-852EA 142.95 USD
100208-852 100204-536
Vanadium(III) acetylacetonate ≥98%
Vanadium(III) acetylacetonate

Metal Beta-diketonates, Volatile Precursors for CVD

Formula: C₁₅H₂₁O₆V
Boiling Pt: 170 °C (0.05 -rr)
Melting Pt: 185 - 190 °C
Density: ~ 1 g/cm³ (20 °C)
MDL Number: MFCD00000033
CAS Number: 13476-99-8
EINECS: 236-759-1
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR