Vanadium ≥99.5%, rods

Supplier: STREM CHEMICALS INC MS

23-0080-10CM 23-0080-2CM
100209-248EA 1917.85 USD
100209-248 100208-850
Vanadium ≥99.5%, rods
Vanadium
Formula: V
MW: 50.94 g/mol
Boiling Pt: 3000 °C
Melting Pt: 1717 °C
Density: 6.015 g/cm³ (20 °C)
MDL Number: MFCD00011453
CAS Number: 7440-62-2
EINECS: 231-171-1
Merck Index: 12,10053

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR