Titanium(IV) chloride ≥99%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Titanium tetrachloride
22-1150-250G
22-1150-1000G
100198-704EA
77.85
USD
100198-704
100200-890
Titanium(IV) chloride ≥99%
Titanium(IV) chloride
Halides for CVD & ALD, Volatile Precursors for CVD
Formula:
Cl₄Ti MW: 189.69 g/mol Boiling Pt: 136 °C (1013 hPa) Melting Pt: 136.2 °C Density: 1.72 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011267 CAS Number: 7550-45-0 EINECS: 231-441-9 UN: 1838 ADR: 8,II Merck Index: 13,09555 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...