Titanium ≥99.7%, foil

Supplier: STREM CHEMICALS INC MS
Warning

22-0151-170MM 22-0151-500MM
100198-148EA 135.88 USD
100198-148 100202-648
Titanium ≥99.7%, foil
Titanium
Formula: Ti
MW: 47.88 g/mol
Boiling Pt: 3262
Melting Pt: 1660
Density: 4.5
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR