Diammonium tetrachloropalladate ≥99.998% (Pd basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Ammonium tetrachloropalladate(II, Ammonium tetrachloropalladate(II), Ammonium palladium (II) chloride, Ammonium tetrachloropalladate (II)

02-1275-1G 02-1275-5G
100197-278EA 167.02 USD
100197-278 100210-770
Diammonium tetrachloropalladate ≥99.998% (Pd basis), PURATREM
Diammonium tetrachloropalladate
Formula: H₈Cl₄N₂Pd
MW: 284.31 g/mol
Melting Pt: Dec.
Density: 2.17 g/cm³ (20 °C)
MDL Number: MFCD00010884
CAS Number: 13820-40-1
EINECS: 237-498-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR