Discandium trioxide ≥99.99% (Sc basis), PURATREM
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Scandium oxide, Scandium (III) oxide
21-2112-5G
21-2112-1G
100200-260EA
271.75
USD
100200-260
100198-698
Discandium trioxide ≥99.99% (Sc basis), PURATREM
Discandium trioxide
Formula:
O₃Sc₂ MW: 137.91 g/mol Melting Pt: 1000 °C Density: 3.86 g/cm³ (20 °C) |
MDL Number:
MFCD00011223 CAS Number: 12060-08-1 EINECS: 235-042-0 Merck Index: 14,08392 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...