Discandium trioxide ≥99.9% (Sc basis), sintered lumps

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Scandium oxide, Scandium (III) oxide

21-0750-1G 21-0750-5G
100206-464EA 50.95 USD
100206-464 100210-794
Discandium trioxide ≥99.9% (Sc basis), sintered lumps
Discandium trioxide
Formula: O₃Sc₂
MW: 137.91 g/mol
Melting Pt: 1000 °C
Density: 3.86 g/cm³ (20 °C)
MDL Number: MFCD00011223
CAS Number: 12060-08-1
EINECS: 235-042-0
Merck Index: 14,08392

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR