Ammonium dihydrogen phosphate ≥99.998%, PURATREM
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Ammonium phosphate, monobasic, Ammonium Phosphate Monobasic, Ammonium dihydrogenphosphate
02-0500-50G
02-0500-10G
100201-158EA
259.02
USD
100201-158
100204-754
Ammonium dihydrogen phosphate ≥99.998%, PURATREM
Ammonium dihydrogen phosphate
Formula:
NH₄H₂PO₄ MW: 115.03 g/mol Melting Pt: 190 °C Density: 1.8 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003396 CAS Number: 7722-76-1 EINECS: 231-764-5 Merck Index: 13,00547 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...