Potassium sulfate ≥99.999% (K basis), PURATREM

Supplier: STREM CHEMICALS INC MS

19-4000-50G 19-4000-10G 19-4000-2G
100203-750EA 649.66 USD
100203-750 100207-610 100207-612
Potassium sulfate ≥99.999% (K basis), PURATREM
Potassium sulphate
Formula: K₂SO₄
Boiling Pt: 1698 °C (1013 hPa)
Melting Pt: 1069 °C
Density: 2.66 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011388
CAS Number: 7778-80-5
EINECS: 231-915-5
Merck Index: 13,07759

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR