Potassium iodide ≥99% ACS

Supplier: STREM CHEMICALS INC MS
Warning

19-3200-500G 19-3200-100G
100209-758EA 184 USD
100209-758 100204-992
Potassium iodide ≥99% ACS
Potassium iodide
Formula: Kl
Boiling Pt: 1330 °C
Melting Pt: 686 °C
Density: 3.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011405
CAS Number: 7681-11-0
EINECS: 231-659-4
Merck Index: 13,07727

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR