Potassium ethanolate ≥95%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Potassium ethylate, Potassium ethoxide
19-1919-100G
19-1919-500G
100206-456EA
111.82
USD
100206-456
100198-690
Potassium ethanolate ≥95%
Potassium ethanolate
Alkoxide
Formula:
C₂H₅KO MW: 84.16 g/mol |
MDL Number:
MFCD00054277 CAS Number: 917-58-8 EINECS: 213-029-0 UN: 3206 ADR: 4.2 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...