Tris(3-methylphenyl)phosphine ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Tris(m-tolyl)phosphine, Tri-m-tolylphosphine
15-8100-5G
15-8100-25G
100207-602EA
84.92
USD
100207-602
100200-874
Tris(3-methylphenyl)phosphine ≥98%
Tris(3-methylphenyl)phosphine
Phosphine
Formula:
C₂₁H₂₁P MW: 304.37 g/mol Melting Pt: 100 °C |
MDL Number:
MFCD00008532 CAS Number: 6224-63-1 EINECS: 228-312-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...