Tris(pentafluorophenyl)phosphine ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Perfluorotriphenylphosphine, Tris(perfluorophenyl)phosphine

15-8005-5G 15-8005-1G
100207-236EA 345.35 USD
100207-236 100198-686
Tris(pentafluorophenyl)phosphine ≥98%
Tris(pentafluorophenyl)phosphine

Phosphine

Formula: C₁₈F₁₅P
MW: 532.15 g/mol
Melting Pt: 117…119 °C
MDL Number: MFCD00079654
CAS Number: 1259-35-4
EINECS: 215-021-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR