Tris(4-methoxyphenyl)phosphine ≥98%
Supplier: STREM CHEMICALS INC MS
Synonyms:
Tris(p-methoxyphenyl)phosphine
15-8000-5G
15-8000-1G
100196-124EA
226.46
USD
100196-124
100209-748
Tris(4-methoxyphenyl)phosphine ≥98%
Tris(4-methoxyphenyl)phosphine
Phosphine
Formula:
C₂₁H₂₁O₃P MW: 352.37 g/mol Boiling Pt: 446 °C (1013 hPa) Melting Pt: 135 °C |
MDL Number:
MFCD00014896 CAS Number: 855-38-9 EINECS: 212-723-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...